missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Neurotrypsin Synthetic Peptide

Product Code. 15296689
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15296689 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15296689 Supplier Invitrogen™ Supplier No. PEP0483
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 23 amino acids near the amino terminus of human Neurotrypsin PEP-0483 can be used as a blocking peptide with polyclonal antibody PA5-20363.

Neurotrypsin is a central nervous system-expressed serine protease whose truncation or absence causes nonsyndromic mental retardation. It is most prominently expressed in structures that are involved in the processing and storage of learned behavior and memory, such as the cerebral cortex, the hippocampus, and amygdala. Evidence suggests that neurotrypsin has multiple functions, including axonal outgrowth, maintaining neuronal plasticity, and arranging the perineuronal environment, partly in coordination with other proteases including tissue plasminogen activator. At least two isoforms of neurotrypsin are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 8492
Species Human
Purification Method purified
Accession Number P56730
Common Name Neurotrypsin
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias Brain-specific serine protease 3; Bssp3; BSSP-3; leydin; Motopsin; MRT1; Neurotrypsin; Nt; peptidase, serine, 12 (neurotrypsin, motopsin); protease, serine 12; protease, serine 12 neurotrypsin (motopsin); protease, serine, 12 (neurotrypsin, motopsin); protease, serine, 12 neurotrypsin (motopsin); Prss12; Serine protease 12
Gene Symbol PRSS12
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.