missing translation for 'onlineSavingsMsg'
Learn More

Human Neurotensin Antibody, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  R&D Systems MAB11548-100

 View more versions of this product

Product Code. 30130388

  • £447.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Neurotensin Monoclonal antibody specifically detects Neurotensin in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Neurotensin
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
neurotensin, neurotensin/neuromedin N, NMN-125, NN, NT, NT/N, NTS, NTS1
Neurotensin containing synthetic peptide, Accession # P30990
100 μg
Primary
Reconstitute lyophilized material at 0.2mg/ml in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a
Immunohistochemistry
1075611
Immunohistochemistry 3-25 μg/mL
P30990
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
4922
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.