missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neuroplastin Isoform 1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5174
This item is not returnable.
View return policy
Description
Neuroplastin Polyclonal specifically detects Neuroplastin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Neuroplastin | |
| Polyclonal | |
| Unconjugated | |
| Q9Y639.1 | |
| NPTN | |
| Mouse myeloma cell line NS0-derived recombinant human Neuroplastin 55 isoform 1 Gln29-Ser219 Accession # Q9Y639.1 | |
| 100 μg | |
| Primary | |
| Detects human Neuroplastin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 1-15 ug/mL | |
| AW554172, GP55, GP55Stromal cell-derived receptor 1, GP65SDR-1, GP66, MGC102805, neuroplastin, np55, Np65, NPTN, SDFR1, SDFR1DKFZp686L2477, SDR1, SDR1stromal cell derived factor receptor 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 27020 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction