missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neuropilin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB22151-SP
This item is not returnable.
View return policy
Description
Neuropilin-2 Monoclonal specifically detects Neuropilin-2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Neuropilin-2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| NP_003863 | |
| NRP2 | |
| Mouse myeloma cell line NS0-derived recombinant human Neuropilin-2 Gln23-Tyr855 Accession # NP_003863 | |
| 25 μg | |
| Primary | |
| Detects human Neuropilin-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat (rr) Neuropilin-1 or rrNeuropilin-2 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 257103 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC126574, neuropilin 2, neuropilin-2, Neuropilin2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8828 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction