missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neuropilin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB38701
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Neuropilin-1 Monoclonal specifically detects Neuropilin-1 in Human samples. It is validated for Western Blot.
Spécification
| Neuropilin-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_001019799 | |
| NRP1 | |
| Mouse myeloma cell line NS0-derived recombinant human Neuropilin-1 Phe22-Lys644 Accession # NP_001019799 | |
| 100 μg | |
| Primary | |
| Detects human Neuropilin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant rat (rr) Neuropilin-1 or rhNeuropilin-2. | |
| Human | |
| Purified |
| Western Blot | |
| 446915 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8829 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu