missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neuropeptide Y/NPY Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £328.00
Specifica
| Antigen | Neuropeptide Y |
|---|---|
| Clone | 904049 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
18162881
![]() |
R&D Systems
MAB85172-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18109290
![]() |
R&D Systems
MAB85172-100 |
100 μg |
£328.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Neuropeptide Y Monoclonal specifically detects Neuropeptide Y in Human samples. It is validated for ELISA.Specifica
| Neuropeptide Y | |
| ELISA | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, NPY, PYY4, seprase | |
| Neuropeptide Y/NPY conjugated to KLH Accession # P01303 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Neuropeptide Y/NPY in direct ELISAs. |
| 904049 | |
| ELISA | |
| Unconjugated | |
| Human | |
| P01303 | |
| 4852 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto