missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Neuroligin 4X/NLGN4X (NP_001269074.1) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | Neuroligin 4X/NLGN4X |
| Accession Number | NP_001269074.1 |
| Gene ID (Entrez) | 57502 |
| Gene Alias | ASPGX2, AUTSX2, HLNX, HNL4X, HNLX, KIAA1260AUTSX2, MGC22376, neuroligin 4, X-linked, NL4, NLGN, NLGN4, NLGN4neuroligin 4, NLGN4X, X-linked |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction