missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neurogenin-3 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF3444-SP
This item is not returnable.
View return policy
Description
Neurogenin-3 Polyclonal specifically detects Neurogenin-3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Neurogenin-3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, NEUROG3, neurogenin 3, neurogenin-3, Neurogenin3, NGN3, Protein atonal homolog 5, Relax | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 50674 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9Y4Z2 | |
| NEUROG3 | |
| E. coli-derived recombinant human Neurogenin-3 Met1-Leu214 Accession # Q9Y4Z2 | |
| 25 μg | |
| Primary | |
| Detects human Neurogenin-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse Neurogenin-3 is observed, and less than 1% cross-reactivity with recombinant human Neurogenin-1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction