missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neurogenin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3524-SP
This item is not returnable.
View return policy
Description
Neurogenin-1 Monoclonal specifically detects Neurogenin-1 in Human samples. It is validated for Western Blot.
Specifications
| Neurogenin-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_006152 | |
| NEUROG1 | |
| E. coli-derived recombinant human Neurogenin-1 Ala145-His237 Accession # NP_006152 | |
| 25 μg | |
| Primary | |
| Detects human Neurogenin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Neurogenin-3 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 309707 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, NEUROG1, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, Neurogenin1, NGN, NGN1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4762 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction