missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neuregulin-1/NRG1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
£171.00 - £1795.00
Specifications
| Antigen | Neuregulin-1/NRG1 |
|---|---|
| Clone | 147724 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18752499
![]() |
R&D Systems
MAB377-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18731448
![]() |
R&D Systems
MAB377-100 |
100 μg |
£454.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18719688
![]() |
R&D Systems
MAB377-500 |
500 μg |
£1795.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neuregulin-1/NRG1 Monoclonal specifically detects Neuregulin-1/NRG1 in Human samples. It is validated for Western Blot.Specifications
| Neuregulin-1/NRG1 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| AAA58639 | |
| 3084 | |
| E. coli-derived recombinant human Neuregulin-1/NRG1 Ser2-Lys246 Accession # AAA58639 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Neuregulin-1/NRG1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with the EGF-like domains of recombinant human (rh) NRG1-beta 1 (aa 176-246) or rhNRG1-alpha (aa 177-241). Therefore, this antibody recognizes an epitope located within aa 2-176 segment N-terminal to the EGF-like domain. This segment is present in all NRG1-alpha , beta and gamma isoforms and in NDF43 and glial growth factor (GGF). It is absent from the NRG1 gene products GGF2 and SMDF. |
| 147724 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARIA, GGF2, GGFglial growth factor, Heregulin-1, HGL, HGLneu differentiation factor, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, Neuregulin1, NRG1, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF | |
| NRG1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title