missing translation for 'onlineSavingsMsg'
Learn More

Human Nestin Antibody, R&D Systems™

Product Code. 18150961
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18150961 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18150961 Supplier R&D Systems Supplier No. MAB1259

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nestin Monoclonal specifically detects Nestin in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nestin
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 196908
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 0.25 μg/106 cells, Immunocytochemistry 8 to 25 μg/mL, CyTOF-reported
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias FLJ21841, NES, nestin
Gene Symbols NES
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Nestin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10763
Test Specificity Detects human Nestin.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.