missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Nesfatin-1 Recombinant Protein, PeproTech®

Product Code. 30130865
Change view
Click to view available options
Quantity:
10 x 100 μg
2 x 100 μg
5 x 100 μg
Unit Size:
Pack of 10
Pack of 2
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30130865 10 x 100 μg Pack of 10
30131512 2 x 100 μg Pack of 2
30132245 5 x 100 μg Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30130865 Supplier Gibco™ Supplier No. 3006701M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

300-67-1MG will be provided as 10 x 100 μg (300-67-100UG). 300-67-200UG will be provided as 2 x 100 μg (300-67-100UG). 300-67-500UG will be provided as 5 x 100 μg (300-67-100UG). Recombinant Human Nesfatin-1 is a 9.7 kDa protein containing 82 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

NUCB2 was initially identified as calcium-binding protein EF hand motif-containing protein that could bind with Necdin, a growth suppressor expressed primarily in postmitotic neurons. NUC2 is a secreted protein that is cleaved into three major peptide products, Nesfatin-1, Nesfatin-2, and Nesfatin-3, of which Nesfatin-1 has been found to cause the suppression of food intake in a leptin-independent manner. Other studies have suggested that NUCB2/Nesfatin-1 may also play roles in energy homeostasis and closely related neuroendocrine functions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P80303
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 4925
Molecular Weight (g/mol) 9.7 kDa
Name Human Nesfatin-1
Quantity 10 x 100 μg
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI607786; Calnuc; DNA-binding protein NEFA; epididymis secretory protein Li 109; Gastric cancer antigen Zg4; HEL-S-109; LOW QUALITY PROTEIN: nucleobindin-2; NEFA; nesfatin 1; nesfatin-1; Nucb2; nucleobindin 2; nucleobindin-2; Nucleobindin-2/Nesfatin-1; nucleobindin-2-like protein; nucleobinding 2; p54; prepronefastin; prepronesfatin
Common Name NUCB2
Biological Activity Determined by an in vivo assay using healthy wild type male mice (C57BL/6 J). Mice were treated via intraperitoneal injection once (at a dose of 4 ug Nesfatin-1/gm of body weight. Significant effects on body weight and food consumption were observed relative to saline-treated controls.)
Conjugate Unconjugated
Recombinant Recombinant
Sequence VPIDIDKTKV QNIHPVESAK IEPPDTGLYY DEYLKQVIDV LETDKHFREK LQKADIEEIK SGRLSKELDL VSHHVRTKLD EL
Content And Storage -20°C
Activity Determined by an in vivo assay using healthy wild type male mice (C57BL/6 J). Mice were treated via intraperitoneal injection once (at a dose of 4 ug Nesfatin-1/gm of body weight. Significant effects on body weight and food consumption were observed relative to saline-treated controls.)
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.