missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nesfatin-1/Nucleobindin-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £362.00
Specifications
| Antigen | Nesfatin-1/Nucleobindin-2 |
|---|---|
| Clone | 877140 |
| Concentration | LYOPH |
| Dilution | Western Blot 3 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18782648
![]() |
R&D Systems
MAB5949-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18081852
|
R&D Systems
MAB5949 |
100 μg |
£362.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nesfatin-1/Nucleobindin-2 Monoclonal specifically detects Nesfatin-1/Nucleobindin-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| Nesfatin-1/Nucleobindin-2 | |
| LYOPH | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| P80303 | |
| 4925 | |
| E. coli-derived recombinant human Nesfatin-1/Nucleobindin-2 Val25-Leu106 Accession # P80303 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Nesfatin-1/Nucleobindin-2 in direct ELISAs and Western blots. |
| 877140 | |
| Western Blot 3 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DNA-binding protein NEFA, NEFA, NEFAGastric cancer antigen Zg4, Nesfatin1, NUCB2, nucleobindin 2, nucleobindin-2, nucleobinding 2 | |
| NUCB2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title