missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neprilysin-2/MMEL1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£146.00 - £345.00
Specifications
| Antigen | Neprilysin-2/MMEL1 |
|---|---|
| Clone | 353207 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18719698
![]() |
R&D Systems
MAB2340-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18108880
![]() |
R&D Systems
MAB2340 |
100 μg |
£345.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neprilysin-2/MMEL1 Monoclonal specifically detects Neprilysin-2/MMEL1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Neprilysin-2/MMEL1 | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q495T6 | |
| 79258 | |
| Mouse myeloma cell line NS0-derived recombinant human Neprilysin-2 Gly78-Trp779 Accession # Q495T6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Neprilysin-2/MMEL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Neprilysin, rhECE-1 or rhECE-2 is observed. |
| 353207 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24, EC 3.4.24.11, MELL1, membrane metallo-endopeptidase-like 1, Membrane metallo-endopeptidase-like 2MGC119455, MGC119454, MMEL1, MMEL2, NEP2, NEP2(m), NEPIIMGC119456, Neprilysin II, neprilysin-2, Neprilysin2, NL2, NL2NL1, SEP, soluble secreted endopeptidase, zinc metallopeptidase | |
| MMEL1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title