missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nephronectin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8025-SP
This item is not returnable.
View return policy
Description
Nephronectin Monoclonal antibody specifically detects Nephronectin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Nephronectin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 1.7-25 ug/mL | |
| Q6UXI9 | |
| NPNT | |
| Chinese hamster ovary cell line CHO-derived recombinant human Nephronectin Glu20-Arg565 Accession # Q6UXI9 | |
| 25 μg | |
| Primary | |
| Detects human Nephronectin in ELISAs and Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 859326 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGFL6L, Nctn, nephronectin, NPNT, POEM | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 255743 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction