missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nephrin Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF4269
This item is not returnable.
View return policy
Description
Nephrin Polyclonal specifically detects Nephrin in Human samples. It is validated for Western Blot.
Specifications
| Nephrin | |
| Polyclonal | |
| Biotin | |
| O60500 | |
| NPHS1 | |
| Mouse myeloma cell line NS0-derived recombinant human Nephrin Gln23-Thr1029 Accession # O60500 | |
| 50 μg | |
| Primary | |
| Detects human Nephrin in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse Nephrin is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CNF, nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, NPHS1, Renal glomerulus-specific cell adhesion receptor | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4868 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction