missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nephrin Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £398.00
Specifications
| Antigen | Nephrin |
|---|---|
| Clone | 1037720 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792357
![]() |
R&D Systems
MAB42692-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18703917
![]() |
R&D Systems
MAB42692-100 |
100 μg |
£398.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nephrin Monoclonal antibody specifically detects Nephrin in Human samples. It is validated for Western BlotSpecifications
| Nephrin | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CNF, nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, NPHS1, Renal glomerulus-specific cell adhesion receptor | |
| Mouse myeloma cell line NS0-derived human Nephrin, Gln23-Thr1029, Accession # O60500 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1037720 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O60500 | |
| 4868 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title