missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NENF Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
NENF Synthetic Peptide, PEP-0697, from Invitrogen.
NENF is a secreted protein that is expressed in neurons but not glial cells of the brain. This protein has neurotrophic activity in primary cultured mouse neurons but not mitogenic activity in primary cultured mouse astrocytes. NENF activated mitogen-activated protein (MAP) and phosphotidylinositol-3 kinase pathways and could be inhibited by pertussis toxin but not receptor tyrosine kinases, suggesting that NENF acts through a Gi/Go-protein-coupled receptor.
Specifications
Specifications
| Gene ID (Entrez) | 29937 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q9UMX5 |
| Common Name | Human NENF |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction