missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NEDD8 (aa 1-36) Control Fragment Recombinant Protein

Product Code. 30195257
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195257

Brand: Invitrogen™ RP108442

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NEDD8 is a ubiquitin-like protein which plays an important role in cell cycle control and embryogenesis. Covalent attachment to its substrates requires prior activation by the E1 complex UBE1C-APPBP1 and linkage to the E2 enzyme UBE2M. Attachment of NEDD8 to cullins activates their associated E3 ubiquitin ligase activity, and thus promotes polyubiquitination and proteasomal degradation of cyclins and other regulatory proteins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15843
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4738
Name Human NEDD8 (aa 1-36) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias nedd8; NEDD-8; NEDD8 ubiquitin like modifier; nedd8.L; nedd8.S; neddylin; Neural precursor cell expressed developmentally down-regulated protein 8; neural precursor cell expressed, developmentally down-regulated 8; neural precursor cell expressed, developmentally down-regulated 8 L homeolog; neural precursor cell expressed, developmentally down-regulated 8 S homeolog; neural precursor cell expressed, developmentally down-regulated gene 8; neural precursor cell-expressed developmentally down-regulated; Rub1; si:zc14a17.5; ubiquitin; ubiquitin-like protein Nedd8; XELAEV_18007009mg
Common Name NEDD8
Gene Symbol Nedd8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MLIKVKTLTGKEIEIDIEPTDKVERIKERVEEKEGI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.