Learn More
ACROBiosystems Human Nectin-2 / CD112 Protein, His Tag
Description
Poliovirus receptor-related 2 (PVRL2) is also known as nectin-2 and CD112 (formerly herpesvirus entry mediator B, HVEB), which is a human plasma membrane glycoprotein. PVRL2 is one of the plasma membrane components of adherens junctions. PVRL2 also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in PVRL2 gene have been associated with differences in the severity of multiple sclerosis. Also, PVRL2 binds with low affinity to TIGIT. Furthermore, PVRL2 acts as a receptor for herpes simplex virus 1 (HHV-1) mutant Rid1, herpes simplex virus 1 (HHV-2) and pseudorabies virus (PRV).
Specifications
Specifications
| Accession Number | NP_002847.1 |
| Molecular Weight (g/mol) | 37.4 kDa |
| Quantity | 100 μg |
| Source | Human Nectin-2, His Tag (PV2-H52E2) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2). |
| Immunogen | Gln 32 - Leu 360 |
| Storage Requirements | -20°C |
| Gene Alias | CD112,Nectin-2,PVRL2 |
| Common Name | Nectin-2 |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.