missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Nectin-2 / CD112 Protein, Fc Tag

Product Code. 15857567
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15857567 100 μg 100µg
15867567 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15857567 Supplier ACROBiosystems Supplier No. PV2H5253100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Poliovirus receptor-related 2 (PVRL2) is also known as nectin-2 and CD112 (formerly herpesvirus entry mediator B, HVEB), which is a human plasma membrane glycoprotein. PVRL2 is one of the plasma membrane components of adherens junctions. PVRL2 also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in PVRL2 gene have been associated with differences in the severity of multiple sclerosis. Also, PVRL2 binds with low affinity to TIGIT. Furthermore, PVRL2 acts as a receptor for herpes simplex virus 1 (HHV-1) mutant Rid1, herpes simplex virus 1 (HHV-2) and pseudorabies virus (PRV).

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.2 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_002847.1
Molecular Weight (g/mol) 62.2 kDa
Quantity 100 μg
Source Human Nectin-2, Fc Tag (PV2-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).
Immunogen Gln 32 - Leu 360
Storage Requirements -20°C
Gene Alias CD112,Nectin-2,PVRL2
Common Name Nectin-2
Conjugate Unconjugated
Species Human
Protein Tag Fc Tag
Buffer Tris with Glycine, Arginine and NaCl, pH7.5
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Immune Checkpoint Proteins
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.