missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Nectin-2 (CD112) Fc Chimera Recombinant Protein
Description
It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1493 μL) be added to the vial to prepare a stock solution of 0.67 mg/mL. Concentration is measured by UV.
This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
| Accession Number | Q92692 |
| Concentration | 0.25 mg/mL |
| For Use With (Application) | Control, ELISA Standard, Binding Assay |
| Format | Lyophilized |
| Formulation | PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4 |
| Gene ID (Entrez) | 5819 |
| Molecular Weight (g/mol) | 70-80 kDa |
| Name | Human Nectin-2 (CD112) Fc Chimera |
| pH Range | 7.4 |
| Purification Method | Purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?