missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Nectin-2/CD112 DuoSet ELISA

Product Code. 30001035
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001035 1 Kit (for 5 Plates) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001035 Supplier R&D Systems™ Supplier No. DY222905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Nectin-2/CD112 DuoSet ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

This DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human Nectin-2/CD112. The suggested diluent is suitable for the analysis of most cell culture supernate samples. Diluents for complex matrices, such as serum and plasma, should be evaluated prior to use in this DuoSet ELISA.

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78.10 to 5000 pg/mL
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 5819
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HVEB, HVEBpoliovirus receptor-like 2, MPH, nectin 2, Nectin2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2, PRR2nectin-2, PVRL2, PVRR2poliovirus receptor related 2
Target Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.