missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NECAP2 (aa 163-253) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55760 (PA5-55760. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q9NVZ3 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 55707 |
| Name | Human NECAP2 (aa 163-253) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1110005F07Rik; AA409105; adaptin ear-binding coat-associated protein 2; adaptin-ear-binding coat-associated protein 2; C78898; clathrin-coated vesicle coat protein; likely ortholog of H. sapiens adaptin-ear-binding coat-associated protein 2 (NECAP2); NECAP endocytosis associated 2; NECAP endocytosis-associated protein 2; Necap2; NECAP-2 |
| Common Name | NECAP2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction