missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NDUFAF5 (aa 164-257) Control Fragment Recombinant Protein

Product Code. 30182607
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182607

Brand: Invitrogen™ RP99326

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62340 (PA5-62340. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The NADH-ubiquinone oxidoreductase complex (complex I) of the mitochondrial respiratory chain catalyzes the transfer of electrons from NADH to ubiquinone, and consists of at least 43 subunits. The complex is located in the inner mitochondrial membrane. This gene encodes a mitochondrial protein that is associated with the matrix face of the mitochondrial inner membrane and is required for complex I assembly. A mutation in this gene results in mitochondrial complex I deficiency. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5TEU4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79133
Name Human NDUFAF5 (aa 164-257) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310003L22Rik; arginine-hydroxylase NDUFAF5, mitochondrial; bA526K24.2; C13H20orf7; C20orf7; C24H20orf7; dJ842G6.1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5; arginine-hydroxylase NDUFAF5, mitochondrial; NADH:ubiquinone oxidoreductase complex assembly factor 5; ndufaf5; probable methyltransferase C20orf7 homolog, mitochondrial; probable methyltransferase C20orf7, mitochondrial; putative methyltransferase C20orf7, mitochondrial-like protein; putative methyltransferase NDUFAF5; RGD1309829; zgc:162919
Common Name NDUFAF5
Gene Symbol NDUFAF5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VNDLPRALEQIHYILKPDGVFIGAMFGGDTLYELRCSLQLAETEREGGFSPHISPFTAVNDLGHLLGRAGFNTLTVDTDEIQVNYPGMFELMED
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado