missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDUFA8 Partial ORF (NP_055037.1, 1 a.a. - 72 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004702-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16004445

  • £280.00 / 10µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the complex I 19 kDA subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It plays an important role in transferring electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [provided by RefSeq]

Sequence: MPGIVELPTLEELKVDEVKISSAVLKAAAHHYGAQCDKPNKEFMLCRWEEKDPRRCLEEGKLVNKCALDFFR
Specifications

Specifications

NP_055037.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.66kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MPGIVELPTLEELKVDEVKISSAVLKAAAHHYGAQCDKPNKEFMLCRWEEKDPRRCLEEGKLVNKCALDFFR
RUO
NDUFA8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4702
NDUFA8 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CI-19KD/CI-PGIV/MGC793/PGIV
NDUFA8
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers