missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NDUFA13 (aa 50-144) Control Fragment Recombinant Protein

Product Code. 30181996
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181996

Brand: Invitrogen™ RP99065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59482 (PA5-59482. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

A novel gene associated with Retinoid-IFN-induced Mortality (GRIM) GRIM-19 gene was identified. Antisense expression of GRIM-19 confers a strong resistance against IFN/RA-induced death by reducing the intracellular levels of GRIM-19 protein. Overexpression of GRIM-19 enhances cell death in response to IFN/RA. GRIM-19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM-19 is a novel cell death-regulatory molecule.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9P0J0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51079
Name Human NDUFA13 (aa 50-144) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2700054G14Rik; AU022060; B16.6; CDA016; Cell d; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CGI-39; CI-B16.6; complex I B16.6 subunit; complex I-B16.6; FLJ58045; FLJ59191; gene associated with retinoic and IFN-induced mortality 19 protein; gene associated with retinoic and interferon-induced mortality 19 protein; Gene associated with retinoic-interferon-induced mortality 19 protein; genes associated with retinoid-IFN-induced mortality 19; GRIM19; GRIM-19; LOC100911483; mitochondrial NADH:ubiquinone oxidoreductase B16.6 subunit; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13-like; NADH:ubiquinone oxidoreductase subunit A13; NADH-ubiquinone oxidoreductase B16.6 subunit; Ndufa13; YjeF N-terminal domain-containing protein 3; YjeF_N3; Yjefn3
Common Name NDUFA13
Gene Symbol Ndufa13
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IMKWNRERRRLQIEDFEARIALLPLLQAETDRRTLQMLRENLEEEAIIMKDVPDWKVGESVFHTTRWVPPLIGELYGLRTTEEALHASHGFMWYT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.