missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NDP (aa 26-133) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82059 (PA5-82059. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
Specifications
Specifications
| Accession Number | Q00604 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 4693 |
| Name | Human NDP (aa 26-133) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | EVR2; FEVR; ND; NDP; NDP, norrin cystine knot growth factor; Ndph; Norrie disease (pseudoglioma); Norrie disease (pseudoglioma) (human); Norrie disease homolog; norrie disease protein; Norrie disease protein homolog; norrin; norrin cystine knot growth factor NDP; RGD1563968; similar to norrin; X-linked exudative vitreoretinopathy 2 protein |
| Common Name | NDP |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction