Learn More
Invitrogen™ Human NCoR2 (aa 2016-2027) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP043
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human SMRT (residues 2016-2027) Synthetic Peptide, PEP-043, from Invitrogen.
PEP-043 is a 15 amino acid synthetic peptide whose sequence corresponds to residues 2016-2027 of human SMRT and an N-terminal addition of C G G as a linker for peptide conjugation to a carrier protein. The sequence of this peptide is (amino to carboxy terminus): C-G-G-A2016-S-A-S-D-P-H-R-E-K-T-Q2027This peptide may be used for neutralization and control experiments with the monoclonal antibody that reacts with this product and SMRT, catalog # MA1-842 & MA1-843. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
9612 | |
purified | |
Human NCoR2 (aa 2016-2027) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q9Y618 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
NCOR2 | |
No preservative | |
50 μg | |
C - G - G - A2016 - S - A - S - D - P - H - R - E - K - T - Q2027 |