missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NCBP2 (aa 132-156) Control Fragment Recombinant Protein

Product Code. 30212055
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212055

Brand: Invitrogen™ RP105232

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66498 (PA5-66498. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The product of this gene is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5' cap of nascent pre-mRNA in the nucleoplasm. The encoded protein has an RNP domain commonly found in RNA binding proteins, and contains the cap-binding activity. The CBC promotes pre-mRNA splicing, 3'-end processing, RNA nuclear export, and nonsense-mediated mRNA decay. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P52298
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 22916
Name Human NCBP2 (aa 132-156) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 20 kDa nuclear cap-binding protein; 20 kDa; 5930413E18Rik; AA536802; AI843301; C79367; CBC2; CBP20; cell proliferation-inducing gene 55 protein; NCBP 20 kDa subunit; NCBP interacting protein 1; NCBP2; NCBP-interacting protein 1; NIP1; nuclear cap binding protein subunit 2; nuclear cap binding protein subunit 2, 20 kD; nuclear cap binding protein subunit 2, 20 kDa; nuclear cap-binding protein subunit 2; phosphatidylinositol glycan anchor biosynthesis, class Z; PIG55; Pigz
Common Name NCBP2
Gene Symbol NCBP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GQVRDEYRQDYDAGRGGYGKLAQNQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.