Learn More
Abnova™ Human NCBP1 Full-length ORF (AAH01450, 1 a.a. - 790 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The product of this gene is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5' cap of nascent pre-mRNA in the nucleoplasm. The encoded protein promotes high-affinity mRNA-cap binding and associates with the CTD of RNA polymerase II. The CBC promotes pre-mRNA splicing, 3'-end processing, RNA nuclear export, and nonsense-mediated mRNA decay. [provided by RefSeq]
Specifications
Specifications
| Accession Number | AAH01450 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 4686 |
| Molecular Weight (g/mol) | 112.64kDa |
| Name | NCBP1 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 10 ug |
| Immunogen | MSRRRHSDENDGGQPHKRRKTSDANETEDHLESLICKVGEKSACSLESNLEGLAGVLEADLPNYKSKILRLLCTVARLLPEKLTIYTTLVGLLNARNYNFGGEFVEAMIRQLKESLKANNYNEAVYLVRFLSDLVNCHVIAAPSMVAMFENFVSVTQEEDVPQVRRDWYVYAFLSSLPWVGKELYEKKDAEMDRIFANTESYLKRRQKTHVPMLQVWTADKPHPQEEYLDCLWAQIQKLKKDRWQERHILRPYLAFDSILCEALQHNLPPFTPPPHTEDSVYPMPRVIFRMFDYTDDPEGPVMPGSHSVERFVIEENLHCIIKSHWKERKTCAAQLVSYPGKNKIPLNYHIVEVIFAELFQLPAPPHIDVMYTTLLIELCKLQPGSLPQVLAQATEMLYMRLDTMNTTCVDRFINWFSHHLSNFQFRWSWEDWSDCLSQDPESPKPKFVREVLEKCMRLSYHQRILDIVPPTFSALCPANPTCIYKYGDESSNSLPGHSVALCLAVAFKSKATNDEIFSILKDVPNPNQDDDDDEGFSFNPLKIEVFVQTLLHLAAKSFSHSFSALAKFHEVFKTLAESDEGKLHVLRVMFEVWRNHPQMIAVLVDKMIRTQIVDCAAVANWIFSSELSRDFTRLFVWEILHSTIRKMNKHVLKIQKELEEAKEKLARQHKRRSDDDDRSSDRKDGVLEEQIERLQEKVESAQSEQKNLFLVIFQRFIMILTEHLVRCETDGTSVLTPWYKNCIERLQQIFLQHHQIIQQYMVTLENLLFTAELDPHILAVFQQFCALQA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.