missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NCAM-1/CD56 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB24083-100
This item is not returnable.
View return policy
Description
NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human samples. It is validated for Immunocytochemistry.
Specifications
| NCAM-1/CD56 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD56, CD56/ NCAM-1, CD56 antigen, MSK39, NCAM1, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| Mouse myeloma cell line NS0-derived recombinant human NCAM-1/CD56, Leu20-Pro603 and Glu636-Asn741, Accession No. NP_001070150 | |
| Primary | |
| Detects human NCAM-1/CD56 in direct ELISAs. | |
| Human | |
| IgG1 |
| Immunocytochemistry | |
| 1006435 | |
| Immunocytochemistry 8-25 μ/mL | |
| NP_001070150 | |
| Mouse | |
| 100 μg | |
| 4684 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction