missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nanog Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1997
This item is not returnable.
View return policy
Description
Nanog Polyclonal specifically detects Nanog in Human samples. It is validated for Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
Specifications
| Nanog | |
| Polyclonal | |
| Unconjugated | |
| ABZ92376 | |
| NANOG | |
| E. coli-derived recombinant human Nanog Trp153-Val305 Accession # ABZ92376 | |
| 50 μg | |
| Primary | |
| Detects human Nanog in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| ChIP Assay, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL | |
| FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 79923 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction