missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NALP5 (aa 399-513) Control Fragment Recombinant Protein

Product Code. 30181643
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181643

Brand: Invitrogen™ RP99156

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (52%), Rat (52%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-144816 (PA5-144816, PA5-63277 (PA5-63277. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP5, also known as MATER, is a maternal gene required for early embryonic development in mice. Increased NALP5 expression was observed in two neuronal injury models, and transient expression of recombinant NALP5 in neurons induced caspase-3 activation and apoptosis, suggesting that NALP5 also plays a role in caspase activation and apoptosis in injured neurons, and may thus represent a novel target for therapeutic treatment in neurodegenerative disorders.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P59047
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 126206
Name Human NALP5 (aa 399-513) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CLR19.8; MATER; Mater protein; Mater protein homolog; maternal antigen that embryos require; NACHT, leucine rich repeat and PYD containing 5; NACHT, LRR and PYD domains-containing protein 5; Nalp5; NLR family pyrin domain containing 5; NLR family, pyrin domain containing 5; NLRP5; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 5; Ooplasm-specific protein 1; OP1; PAN11; PYPAF8
Common Name NALP5
Gene Symbol NLRP5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLPESFLIVTVRDVGTEKLKSEVVSPRYLLVRGISGEQRIHLLLERGIGEHQKTQGLRAIMNNRELLDQCQVPAVGSLICVALQLQDVVGESVAPFNQTLTGLHAAFVFHQLTPR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.