missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NALP4 (aa 701-792) Control Fragment Recombinant Protein

Product Code. 30210331
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210331

Brand: Invitrogen™ RP109015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (47%), Rat (47%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139855 (PA5-139855. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP4, also known as PYRIN-containing APAF1-like protein 4, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. In transfected 293 cells, NALP4 suppressed NF-kappa-B induction by TNF-a and IL-1-b, suggesting NALP4 operates at a point of convergence in these two signaling pathways. NALP4 also suppressed NF-kappa-B induction resulting from overexpression of several adapter proteins and protein kinases involved in these pathways, including TRAF2, TRAF6, RIP, and IRAK2, as well as the IKK-alpha and IKK-beta, suggesting that NALP4 is critical in modulating NF-kappa-B activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96MN2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 147945
Name Human NALP4 (aa 701-792) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4930406H16Rik; cancer/testis antigen 58; CLR19.5; CT58; E330021B02; E330028A19Rik; NACHT, leucine rich repeat and PYD containing 4; NACHT, leucine rich repeat and PYD containing 4 A; NACHT, leucine rich repeat and PYD containing 4 E; NACHT, LRR and PYD containing protein 4; NACHT, LRR and PYD domains-containing protein 4; NACHT, LRR and PYD domains-containing protein 4 A; NACHT, LRR and PYD domains-containing protein 4 E; NACHT/LRR/pyrin domain-containing protein 4; NALP, epsilon; NALP4; Nalp4a; Nalp4e; NALP-epsilon; Nalp-eta; Nalp-ita; NLR family pyrin domain containing 4; NLR family, pyrin domain containing 4; NLR family, pyrin domain containing 4 A; NLR family, pyrin domain containing 4 E; Nlrp4; Nlrp4a; Nlrp4e; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 4; PAAD and NACHT-containing protein 2; PAN2; PYPAF4; PYRIN and NACHT-containing protein 2; PYRIN-containing APAF1-like protein 4; RGD1561918; RGD1563529; Ribonuclease inhibitor 2; RNH2
Common Name NALP4
Gene Symbol NLRP4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FTLTKLSRDDIRSLCDALNYPAGNVKELALVNCHLSPIDCEVLAGLLTNNKKLTYLNVSCNQLDTGVPLLCEALCSPDTVLVYLMLAFCHLS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.