missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NALP12 (aa 654-727) Control Fragment Recombinant Protein

Product Code. 30182376
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182376

Brand: Invitrogen™ RP99104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (54%), Rat (54%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60185 (PA5-60185. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP12, also known as Monarch-1, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. NALP12 is thought to act as an attenuating factor of inflammation by suppressing inflammatory responses such as NF-kappa-B activation by TLR-signaling molecules MyD88, IRAK-1, TRAF6 and RIPK1 in activated monocytes. Recent evidence suggests that mutations in NALP12 result in hereditary periodic fever syndromes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P59046
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 91662
Name Human NALP12 (aa 654-727) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CLR19.3; FCAS2; monarch 1; monarch-1; NACHT, leucine rich repeat and PYD containing 12; NACHT, LRR and PYD containing protein 12; NACHT, LRR and PYD domains-containing protein 12; NALP12; NLR family pyrin domain containing 12; NLR family, pyrin domain containing 12; Nlrp12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 12; PAN6; PYPAF7; PYRIN-containing APAF1-like protein 7; regulated by nitric oxide; RNO; RNO2
Common Name NALP12
Gene Symbol NLRP12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LKRCRSAQVLHLYGATYSADGEDRARCSAGAHTLLVQLPERTVLLDAYSEHLAAALCTNPNLIELSLYRNALGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.