missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NAALADase-like 1/NAALADL1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5467-SP
This item is not returnable.
View return policy
Description
NAALADase-like 1/NAALADL1 Polyclonal specifically detects NAALADase-like 1/NAALADL1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| NAALADase-like 1/NAALADL1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| I100, ileal dipeptidylpeptidase, NAALADase L, NAALADase like 1, NAALADASEL, NAALADase-like 1, NAALADL1, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10004 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| NP_005459 | |
| NAALADL1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human NAALADase-like 1/NAALADL1 Ala32-Leu740 Accession # NP_005459 | |
| 25 μg | |
| Primary | |
| Detects human NAALADase-like 1/NAALADL1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) NAALADase I, recombinant mouse NAALADase II, and rhNAALADase II is observed. | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering