missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NAA40 (aa 75-146) Control Fragment Recombinant Protein

Product Code. 30196067
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196067

Brand: Invitrogen™ RP97302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58121 (PA5-58121. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

N-terminal acetylation is one of the most common protein modifications in eukaryotes, occurring on approximately 57% and 84% on yeast and human proteins respectively. There are several N-terminal acetylating enzyme complexes (NatA - NatE). Unlike the other complexes, NatD is composed of a single protein, NAT11, and has recently been described to acetylate the Serine N-termini of histones H2A and H4 in yeast. The role these modifications play is unknown; yeast that do not express NAT11 grow at normal rates and have no observable phenotypes. The role of the human homolog is likewise unknown.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86UY6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79829
Name Human NAA40 (aa 75-146) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4931433E08Rik; AU023197; N(alpha)-acetyltransferase 40, NatD catalytic subunit; N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog; N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae); Naa40; N-acetyltransferase 11; N-acetyltransferase 11 (GCN5-related, putative); N-alpha acetyl transferase 40; Nalpha acetyltransferase 40; N-alpha-acetyltransferase 40; N-alpha-acetyltransferase 40, NatD catalytic subunit; N-alpha-acetyltransferase D; NAT11; NatD; natD catalytic subunit; PATT1; Protein acetyltransferase 1
Common Name NAA40
Gene Symbol NAA40
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DLTKTNMQTMYEQSEWGWKDREKREEMTDDRAWYLIAWENSSVPVAFSHFRFDVECGDEVLYCYEVQLESKV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.