missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human N6AMT1 (aa 148-212) Control Fragment Recombinant Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66242 (PA5-66242. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on 'Gln-185', using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.
Spezifikation
Spezifikation
| Accession Number | Q9Y5N5 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 29104 |
| Name | Human N6AMT1 (aa 148-212) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 5830445C04Rik; C21orf127; HemK methyltransferase family member 2; HEMK2; m.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase N6AMT1; methyltransferase N6AMT1; hemK methyltransferase family member 2; M-N6DNA MTase A; MTQ2; N(6)-adenine-specific DNA methyltransferase 1; N(6)-adenine-specific DNA methyltransferase 1-like; N-6 adenine-specific DNA methyltransferase 1; N-6 adenine-specific DNA methyltransferase 1 (putative); N6AMT; N6amt1; N6-DNA methyltransferase A; N6-DNA-methyltransferase; PRED28; Protein N(5)-glutamine methyltransferase; RGD1311843 |
| Common Name | N6AMT1 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur