missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human N-Terminal Pro-BNP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9769-SP
This item is not returnable.
View return policy
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for ELISA.
Specifications
| BNP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BNF, BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein, NPPB | |
| E. coli-derived recombinant human Pro-BNP, His27-His134, Accession No. P16860 | |
| 25 μg | |
| 4879.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| ELISA | |
| 817130R | |
| ELISA | |
| P16860 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Pro-BNP but does not detect mature human BNP peptide in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction