missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human N-Cadherin DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human N-Cadherin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human N-Cadherin.
Specifications
Specifications
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Binding Target | N-Cadherin |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1000 |
| Synonym | cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDH2, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction