missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MZB1 (aa 28-94) Control Fragment Recombinant Protein

Product Code. 30193826
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193826

Brand: Invitrogen™ RP104951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84122 (PA5-84122. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MZB1 associates with immunoglobulin M (IgM) heavy and light chains and promotes IgM assembly and secretion. May exert its effect by acting as a molecular chaperone or as an oxidoreductase as it displays a low level of oxidoreductase activity. Isoform 2 may be involved in regulation of apoptosis. Helps to diversify peripheral B-cell functions by regulating Ca(2+) stores, antibody secretion and integrin activation.; Acts as a hormone-regulated adipokine/proinflammatory cytokine that is implicated in causing chronic inflammation, affecting cellular expansion and blunting insulin response in adipocytes. May have a role in the onset of insulin resistance.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WU39
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51237
Name Human MZB1 (aa 28-94) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias caspase-2 binding protein; HSPC190; marginal zone B and B1 cell specific protein; marginal zone B and B1 cell-specific protein; marginal zone B- and B1-cell-specific protein; MEDA7; MEDA-7; Mesenteric estrogen-dependent adipose 7; mesenteric oestrogen-dependent adipose gene- 7; MGC29506; MZB1; PACAP; pERp1; plasma cell-induced ER protein 1; plasma cell-induced resident endoplasmic reticulum protein; Plasma cell-induced resident ER protein; proapoptotic caspase adapter protein; proapoptotic caspase adaptor protein
Common Name MZB1
Gene Symbol MZB1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TATAPQLDDEEMYSAHMPAHLRCDACRAVAYQMWQNLAKAETKLHTSNSGGRRELSELVYTDVLDRS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.