missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Myosin Heavy Chain APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC4470A
This item is not returnable.
View return policy
Description
Myosin Heavy Chain Monoclonal specifically detects Myosin Heavy Chain in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Myosin Heavy Chain | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| MYH14 | |
| Chicken pectoralis-derived Myosin | |
| 100 Tests | |
| Primary | |
| Detects Myosin Heavy Chain in human, mouse, rat and other mammalian, avian, and amphibian species. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| MF20 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| DFNA4, FLJ13881, FLJ43092, KIAA2034DKFZp667A1311, MHC16, MYH14 variant protein, MYH17, MYHC, myosin, Myosin heavy chain 14, Myosin heavy chain, non-muscle IIc, myosin, heavy chain 14, myosin, heavy chain 14, non-muscle, myosin, heavy polypeptide 14, myosin-14, NMHC II-C, NMHC-II-C, nonmuscle myosin heavy chain II-C, Non-muscle myosin heavy chain IIc | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79784 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction