missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Myoglobin Antibody, R&D Systems™
Mouse Monoclonal Antibody
£157.00 - £355.00
Specifications
| Antigen | Myoglobin |
|---|---|
| Clone | 974145 |
| Dilution | Western Blot 0.05μg/mL, Simple Western 10μg/mL, ELISA, Immunohistochemistry 1-25μg/mL |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18151841
|
R&D Systems
MAB97202-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109770
|
R&D Systems
MAB97202-100 |
100 μg |
£355.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myoglobin Monoclonal specifically detects Myoglobin in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry.Specifications
| Myoglobin | |
| Western Blot 0.05μg/mL, Simple Western 10μg/mL, ELISA, Immunohistochemistry 1-25μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P02144 | |
| 4151 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 974145 | |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| MB, MGC13548, myoglobin, PVALB | |
| Purified human cardiac myoglobin antigen from human heart Accession # P02144 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human Myoglobin in direct ELISAs. In sandwich immunoassays, the antibody pair is specific for human Myoglobin. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title