missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MYOC (aa 144-228) Control Fragment Recombinant Protein

Product Code. 30199691
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199691

Brand: Invitrogen™ RP93985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55590 (PA5-55590. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Myocilin is an extracellular protein expressed in the eye, including the retina, trabecular meshwork and ciliary body. Myocilin can form homomultimers in vivo and can also associate with components of the ECM via interactions with the Hep II domain of FibroPVRL1. In addition, myocilin interacts with myosin regulatory light chain, a component of the myosin motor protein complex. This interaction implies a role for myocilin in the actomyosin system, linking myocilin to the functional status of the trabecular meshwork (TM), which is responsible for controlling the intraocular pressure (IOP). Alterations in functions of the TM may lead to IOP elevation and development of glaucoma, a major cause of blindness. Myocilin is encoded by MYOC (also designated TIGR), a gene that maps to the GLC1A locus on chromosome 1q24. 3 and is susceptible to mutations. Mutations in the MYOC gene are specifically linked with primary open angle glaucoma (POAG), a blinding disease characterized by progressive loss of retinal ganglion cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99972
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4653
Name Human MYOC (aa 144-228) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI957332; GLC1A; GPOA; JOAG; JOAG1; juvenile-onset open-angle glaucoma 1; mutated trabecular meshwork-induced glucocorticoid response protein; MYOC; Myocilin; Myocilin 20 kDa N-terminal fragment; Myocilin 35 kDa N-terminal fragment; Myocilin 55 kDa subunit; myocilin trabecular meshwork inducible glucocorticoid response; myocilin trabecular meshwork inducible glucocorticoid response protein; Myocilin, C-terminal fragment; Myocilin, N-terminal fragment; myocilin, trabecular meshwork inducible glucocorticoid response; TIGR; trabecular meshwork inducible glucocorticoid response protein; trabecular meshwork-induced glucocorticoid response protein
Common Name MYOC
Gene Symbol MYOC
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NLLRDKSVLEEEKKRLRQENENLARRLESSSQEVARLRRGQCPQTRDTARAVPPGSREVSTWNLDTLAFQELKSELTEVPASRIL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.