missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MYLPF (aa 112-143) Control Fragment Recombinant Protein

Product Code. 30211839
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211839

Brand: Invitrogen™ RP108209

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84234 (PA5-84234. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MYLPF (myosin light chain, phosphorylatable, fast skeletal muscle), also known as fast skeletal myosin light chain 2 or MLC2B, is a 169 amino acid protein that is expressed in fetal and adult skeletal muscle. A calcium binding protein, MYLPF contains three EF hand domains and is encoded by a gene that maps to human chromosome 16p11.2. Chromosome 16 encodes over 900 genes in approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96A32
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 29895
Name Human MYLPF (aa 112-143) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2410014J02Rik; DKFZp779C0757; DTNB; fast skeletal myosin light chain 2; G2; HUMMLC2B; MGC13450; MLC2; MLC-2; MLC2B; MLC2F; MRLC2; MYL11; Myl2; MYLPF; Myolc1; myosin light chain; myosin light chain 2; myosin light chain 2 type 2; myosin light chain 2 type I; myosin light chain, phosphorylatable, fast skeletal muscle; Myosin light polypeptide 2 alkali; myosin regulatory light chain 2, skeletal muscle isoform; Myosin regulatory light chain 2, skeletal muscle isoform type 2; myosin, light chain 11, regulatory; Myosin, light polypeptide 2, alkali; Myosin, light polypeptide 2, alkali; ventricular, skeletal, slow; phosphorylatable fast skeletal muscle myosin light chain; Unknown (protein for MGC:143425); ventricular skeletal slow; ventricular, skeletal, slow
Common Name MYLPF
Gene Symbol MYLPF
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KGTIKKKFLEELLTTQCDRFSQEEIKNMWAAF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.