missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MYL2 (aa 75-163) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54073 (PA5-54073. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.
Specifications
Specifications
| Accession Number | P10916 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 4633 |
| Name | Human MYL2 (aa 75-163) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | cardiac myosin light chain 2; cardiac myosin light chain-2; cardiac ventricular myosin light chain 2; CMH10; DKFZp779C0562; hypothetical protein LOC677750; Mlc2; MLC-2; mlc2b; MLC-2 s/v; MLC2V; MLC-2 V; myl2; myl2b; Mylpc; myosin light chain 2; myosin light chain 2, slow skeletal/ventricular muscle isoform; myosin light chain 2 V; myosin light chain phosphatase; myosin light chain, phosphorylatable, cardiac ventricles; myosin light chain-2; myosin regulatory light chain 2, ventricular/cardiac muscle isoform; myosin regulatory light chain 2 b; myosin regulatory light chain ventricular isoform; myosin, light chain 2, regulatory, cardiac, slow; myosin, light chain 2 b, regulatory, cardiac, slow; myosin, light polypeptide 2, regulatory, cardiac, slow; myosin, light polypeptide 2 b, regulatory, cardiac, slow; regulatory light chain of myosin; RLC of myosin; slow cardiac myosin regulatory light chain 2; ventricular myosin light chain 2; ventricular myosin regulatory light chain; zgc:136848 |
| Common Name | MYL2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction