missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Myeloperoxidase/MPO Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £423.00
Specifications
| Antigen | Myeloperoxidase/MPO |
|---|---|
| Clone | 358621 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713367
![]() |
R&D Systems
MAB31741-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18723137
![]() |
R&D Systems
MAB31741-100 |
100 μg |
£423.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for ELISASpecifications
| Myeloperoxidase/MPO | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 1.11.1, EC 1.11.1.7, Lactoperoxidase, MPO, myeloperoxidase | |
| Mouse myeloma cell line NS0-derived human Myeloperoxidase/MPO protein, Ala49-Ser745, Accession # P05164 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 358621 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P05164 | |
| 4353 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title