missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human MyD88 Synthetic Peptide
missing translation for 'orderingAttributeHoverText'
Quantity:
50 μg
Förpackningsstorlek
50µg
Beskrivelse
MyD88 Synthetic Peptide, PEP-0044, from Invitrogen.
Myeloid differentiation Marker 88 (MyD88) is an adaptor molecule for the Toll/IL-1R and is involved in the inflammatory response induced by IL-1, IL-18 and LPS. MyD88 recruits IRAK to IL-1 receptor complex in response to IL-1. This pathway further leads to activation of NF-kB. MyD88 has a wide tissue distribution.
Tekniske data
Tekniske data
| Gene ID (Entrez) | 4615 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q99836 |
| Common Name | Human MyD88 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion