missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MYCL1/L-Myc Antibody, R&D Systems™

Mouse Monoclonal Antibody
£143.00 - £387.00
Specifications
| Antigen | MYCL1/L-Myc |
|---|---|
| Clone | 803013 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18750557
![]() |
R&D Systems
MAB4050-SP |
25 μg |
£143.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18741327
![]() |
R&D Systems
MAB4050-100 |
100 μg |
£387.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYCL1/L-Myc Monoclonal specifically detects MYCL1/L-Myc in Human samples. It is validated for ELISA.Specifications
| MYCL1/L-Myc | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P12524 | |
| 4610 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 803013 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BHLHE38, bHLHe38myc-related gene from lung cancer, Class E basic helix-loop-helix protein 38, LMyc, L-Myc, LMYCl-myc-1 proto-oncogene, MYCL, protein L-Myc-1, v-myc avian myelocytomatosis viral oncogene homolog 1, lung carcinoma derived, v-myc myelocytomatosis viral oncogene homolog 1, lung carcinoma derived (avian) | |
| E. coli-derived recombinant human MYCL1/L-Myc, Gly16-Asn139, Accession # P12524 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MYCL1/L-MYC in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title